siRNA Details
| Gene Symbol: Strip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153563.2 | NP_705791.2 | 19 | 2026 | ||
| GenBank mRNA | AK032065.1 | - | 19 | 2008 | ||
| AK122553.1 | - | 17 | 3787 | |||
| AK148011.1 | - | 19 | 2011 | |||
| AK169644.1 | - | 17 | 2574 | |||
| AK188013.1 | - | 4 | 318 | |||
| AK189280.1 | - | 4 | 291 | |||
| BC023952.1 | - | 19 | 2002 | 5320305 | ||

Strip1
Genomic Map