siRNA Details
| Gene Symbol: Rbm41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172147.1 | NP_001165618.1 | 8 | 2367 | ||
| NM_001290630.1 | NP_001277559.1 | 8 | 2357 | |||
| NM_153586.2 | NP_705814.1 | 7 | 2285 | |||
| GenBank mRNA | AK084514.1 | - | 7 | 2221 | ||
| AK084629.1 | - | 7 | 2221 | |||
| AK141075.1 | - | 8 | 2292 | |||
| AK163147.1 | - | 6 | 2205 | |||
| AK164496.1 | - | 8 | 2319 | |||
| BC037024.1 | - | 7 | 2252 | 5368185 | ||

Rbm41
Genomic Map