siRNA Details
| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153782.1 | NP_722477.1 | 10 | 1869 | ||
| GenBank mRNA | AK142932.1 | - | 10 | 1894 | ||
| BC011274.1 | - | 10 | 1773 | |||
| BC023310.1 | - | 3 | 170 | |||
| BC029169.1 | - | 10 | 1869 | 4947019 | ||

Fam20a
Genomic Map