siRNA Details
| Gene Symbol: Flywch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153791.2 | NP_722486.2 | 4 | 1193 | ||
| GenBank mRNA | AK034869.1 | - | 3 | 787 | ||
| AK052314.1 | - | 2 | 405 | |||
| AK129391.1 | - | 2 | 351 | |||
| AK168389.1 | - | 4 | 1128 | |||
| AK197265.1 | - | 1 | 20 | |||
| BC038031.1 | - | 3 | 1485 | 4021754 | ||

Flywch1
Genomic Map