siRNA Details
| Gene Symbol: Spata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170756.2 | NP_739562.1 | 3 | 3096 | ||
| GenBank mRNA | AK122364.1 | - | 1 | 3738 | ||
| AK156188.1 | - | 3 | 3097 | |||
| AK162743.1 | - | 3 | 3096 | |||
| AK171664.1 | - | 3 | 3074 | |||
| AK171852.1 | - | 3 | 3086 | |||
| BC034597.1 | - | 3 | 3085 | 5363513 | ||

Spata2
Genomic Map