siRNA Details
| Gene Symbol: Ccdc186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170757.1 | NP_739563.1 | 5, 6 | 1286 | ||
| GenBank mRNA | AF349751.1 | - | 5, 6 | 1286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc186| Gene Symbol: Ccdc186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170757.1 | NP_739563.1 | 5, 6 | 1286 | ||
| GenBank mRNA | AF349751.1 | - | 5, 6 | 1286 | ||