siRNA Details
| Gene Symbol: Zfp628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170759.2 | NP_739565.2 | 3 | 3157 | ||
| GenBank mRNA | AF435832.1 | - | 1 | 2861 | ||
| AK155214.1 | - | 3 | 3157 | |||
| BC056945.1 | - | 4 | 1964 | |||
| BC062973.1 | - | 4 | 1964 | |||

Zfp628
Genomic Map