siRNA Details
| Gene Symbol: Pddc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172116.4 | NP_742114.3 | 3 | 215 | ||
| GenBank mRNA | AK081187.1 | - | 3 | 175 | ||
| AK084273.1 | - | 3 | 171 | |||
| AK088159.1 | - | 3 | 191 | |||
| AK140975.1 | - | 3 | 166 | |||
| AK161205.1 | - | 3 | 181 | |||
| BC023835.1 | - | 3 | 185 | 5345186 | ||
| BY118625.1 | - | 3 | 215 | |||

Pddc1
Genomic Map