siRNA Details
| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172255.3 | NP_758459.2 | 3 | 335 | ||
| GenBank mRNA | AK170058.1 | - | 3 | 283 | ||
| BC037177.1 | - | 3 | 272 | 4948388 | ||
| CK638976.1 | - | 3 | 324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr11| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172255.3 | NP_758459.2 | 3 | 335 | ||
| GenBank mRNA | AK170058.1 | - | 3 | 283 | ||
| BC037177.1 | - | 3 | 272 | 4948388 | ||
| CK638976.1 | - | 3 | 324 | |||