siRNA Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289668.1 | NP_001276597.1 | 2 | 612 | ||
| NM_172257.4 | NP_758461.1 | 2 | 612 | |||
| GenBank mRNA | AK081177.1 | - | 2 | 598 | ||
| AK152371.1 | - | 2 | 545 | |||
| AK155875.1 | - | 2 | 516 | |||
| AK196459.1 | - | 2 | 144 | |||
| BC023957.1 | - | 2 | 616 | 5321190 | ||
| BC051101.1 | - | 2 | 340 | 5052849 | ||
| CB247121.1 | - | 2 | 621 | |||

Sidt2
Genomic Map