siRNA Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172281.2 | NP_758485.2 | 2, 3 | 329 | ||
| GenBank mRNA | AK089805.1 | - | 2, 3 | 261 | ||
| AK129184.1 | - | 2, 3 | 247 | |||
| AK154982.1 | - | 2, 3 | 260 | |||
| AK171462.1 | - | 2, 3 | 258 | |||
| BC038348.1 | - | 2, 3 | 243 | 4187484 | ||
| BY301442.1 | - | 2, 3 | 332 | |||

Rnf40
Genomic Map