siRNA Details
| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172283.3 | NP_758487.2 | 7, 8 | 680 | ||
| GenBank mRNA | AJ297482.4 | - | 7, 8 | 651 | ||
| AJ534942.1 | - | 7, 8 | 651 | |||
| AK164340.1 | - | 7, 8 | 649 | |||
| AY223865.1 | - | 7, 8 | 651 | |||
| BC037698.1 | - | 6, 7 | 629 | 4190449 | ||

Fuk
Genomic Map