siRNA Details
| Gene Symbol: 6430548M08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163760.1 | NP_001157232.1 | 6 | 904 | ||
| NM_001163761.1 | NP_001157233.1 | 6 | 925 | |||
| NM_001163762.1 | NP_001157234.1 | 6 | 925 | |||
| NM_172286.4 | NP_758490.3 | 6 | 925 | |||
| GenBank mRNA | AK032447.1 | - | 6 | 926 | ||
| AK044754.1 | - | 5 | 734 | |||
| AK046401.1 | - | 6 | 927 | |||
| AK051096.1 | - | 6 | 927 | |||
| AK122299.1 | - | 6 | 911 | |||
| AK133157.1 | - | 6 | 923 | |||
| AK172108.1 | - | 6 | 926 | |||
| BC027126.1 | - | 6 | 904 | 5359842 | ||

6430548M08Rik
Genomic Map