siRNA Details
| Gene Symbol: Nup133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172288.2 | NP_758492.2 | 4 | 587 | ||
| GenBank mRNA | AK029317.1 | - | 4 | 571 | ||
| AK040528.1 | - | 4 | 578 | |||
| AK188122.1 | - | 4 | 342 | |||
| BC023915.1 | - | 4 | 598 | 5323157 | ||
| BC026845.1 | - | 4 | 570 | 4235408 | ||
| BC033476.1 | - | 4 | 584 | |||

Nup133
Genomic Map