siRNA Details
| Gene Symbol: Foxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291448.1 | NP_001278377.1 | 1, 2 | 242 | ||
| NM_001291449.1 | NP_001278378.1 | 1, 2 | 242 | |||
| NM_172291.2 | NP_758495.1 | 1, 2 | 242 | |||
| GenBank mRNA | AK054231.1 | - | 1, 2 | 134 | ||
| AK144759.1 | - | 1, 2 | 168 | |||
| AK163722.1 | - | 1, 2 | 101 | |||
| BC017604.1 | - | 1, 2 | 195 | |||
| BC024806.1 | - | 1, 2 | 112 | 5361053 | ||
| BC027386.1 | - | 1, 2 | 141 | |||

Foxred1
Genomic Map