siRNA Details
| Gene Symbol: Klhl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122830.1 | NP_001116302.1 | 2 | 1512 | ||
| NM_178771.3 | NP_848886.1 | 3 | 1695 | |||
| GenBank mRNA | AK036321.1 | - | 3 | 1537 | ||
| AK041089.1 | - | 3 | 1698 | |||
| AK048601.1 | - | 3 | 1663 | |||
| AK049946.1 | - | 3 | 1695 | |||
| AK163170.1 | - | 3 | 1695 | |||
| BC023909.1 | - | 3 | 1704 | 5322205 | ||
| BC025484.1 | - | 3 | 1606 | 5151231 | ||
| BC098185.1 | - | 3 | 1670 | 3594528 | ||

Klhl26
Genomic Map