siRNA Details
| Gene Symbol: Dio3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172119.2 | NP_742117.2 | 1 | 1060 | ||
| GenBank mRNA | AK140797.1 | - | 1 | 1061 | ||
| AK146060.1 | - | 1 | 1061 | |||
| BC106847.1 | - | 1 | 1030 | 40046057 | ||
| BC106848.1 | - | 1 | 1030 | 40046058 | ||

Dio3
Genomic Map