siRNA Details
| Gene Symbol: Crocc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145958.1 | NP_001139430.1 | 5, 6 | 512 | ||
| NM_172122.2 | NP_742120.2 | 6, 7 | 765 | |||
| GenBank mRNA | AF527975.1 | - | 6, 7 | 765 | ||
| BC054054.1 | - | 4, 5 | 493 | 6394697 | ||
| BQ937768.1 | - | 3, 4 | 291 | |||
| CF731932.1 | - | 4, 5 | 401 | |||

Crocc
Genomic Map