siRNA Details
| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY115494.1 | - | 1 | 195 | ||
| AY444556.1 | - | 1 | 195 | |||
| BC092350.1 | - | 1 | 185 | |||
| BC139385.1 | - | 1 | 194 | 8862407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbsn| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY115494.1 | - | 1 | 195 | ||
| AY444556.1 | - | 1 | 195 | |||
| BC092350.1 | - | 1 | 185 | |||
| BC139385.1 | - | 1 | 194 | 8862407 | ||