siRNA Details
| Gene Symbol: Mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172252.3 | NP_758456.2 | 3, 4 | 279 | ||
| GenBank mRNA | AK003285.1 | - | 3, 4 | 266 | ||
| AK012784.1 | - | 3, 4 | 271 | |||
| AK020739.1 | - | 3, 4 | 280 | |||
| BC028768.1 | - | 3, 4 | 254 | 1282313 | ||

Mrpl21
Genomic Map