siRNA Details
| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172255.3 | NP_758459.2 | 5 | 663 | ||
| GenBank mRNA | AK043051.1 | - | 3 | 281 | ||
| AK166369.1 | - | 5 | 618 | |||
| AK170058.1 | - | 5 | 611 | |||
| BC037177.1 | - | 5 | 600 | 4948388 | ||
| CK638976.1 | - | 5 | 652 | |||

Wdr11
Genomic Map