siRNA Details
| Gene Symbol: Mab21l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172295.4 | NP_758499.3 | 3 | 621 | ||
| GenBank mRNA | AK029040.1 | - | 3 | 360 | ||
| AK161057.1 | - | 3 | 538 | |||
| AK166928.1 | - | 371 | ||||
| AK172389.1 | - | 3 | 570 | |||
| BC037703.1 | - | 3 | 558 | 4166596 | ||

Mab21l3
Genomic Map