siRNA Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172134.2 | NP_742146.1 | 3 | 319 | ||
| GenBank mRNA | AK039194.1 | - | 3 | 301 | ||
| AK080846.1 | - | 3 | 295 | |||
| AK145470.1 | - | 3 | 283 | |||
| AK166078.1 | - | 3 | 302 | |||
| AK166464.1 | - | 3 | 284 | |||
| BC027745.1 | - | 3 | 356 | 5064695 | ||
| BC032171.1 | - | 3 | 290 | |||
| BI143426.1 | - | 3 | 348 | |||

Pdxk
Genomic Map