siRNA Details
| Gene Symbol: Dync2li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172256.1 | NP_758460.1 | 13 | 1187 | ||
| GenBank mRNA | AK008822.1 | - | 12 | 1137 | ||
| AK160259.1 | - | 13 | 1204 | |||
| BC030450.1 | - | 13 | 1189 | 5400492 | ||
| BC039070.1 | - | 13 | 1187 | 3482503 | ||

Dync2li1
Genomic Map