siRNA Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289668.1 | NP_001276597.1 | 10 | 1319 | ||
| NM_172257.4 | NP_758461.1 | 10 | 1319 | |||
| GenBank mRNA | AK152371.1 | - | 10 | 1252 | ||
| AK155875.1 | - | 10 | 1223 | |||
| BC023957.1 | - | 10 | 1323 | 5321190 | ||
| BC051101.1 | - | 10 | 1047 | 5052849 | ||
| BQ179223.1 | - | 3 | 158 | |||

Sidt2
Genomic Map