siRNA Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172281.2 | NP_758485.2 | 20 | 3906 | ||
| GenBank mRNA | AK042654.1 | - | 11 | 2566 | ||
| AK089805.1 | - | 20 | 3837 | |||
| AK129184.1 | - | 20 | 3824 | |||
| AK154982.1 | - | 20 | 3838 | |||
| AK171462.1 | - | 20 | 3836 | |||
| AK207184.1 | - | 1 | 357 | |||
| BC038348.1 | - | 20 | 3820 | 4187484 | ||

Rnf40
Genomic Map