siRNA Details
| Gene Symbol: Slc36a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172289.4 | NP_758493.2 | 9 | 1025 | ||
| GenBank mRNA | AF453746.1 | - | 10 | 1024 | ||
| AK032074.1 | - | 9 | 1026 | |||
| AK052277.1 | - | 9 | 1026 | |||
| AK150167.1 | - | 9 | 1057 | |||
| BC115964.1 | - | 9 | 1026 | 40057255 | ||

Slc36a4
Genomic Map