siRNA Details
| Gene Symbol: Foxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291448.1 | NP_001278377.1 | 2 | 271 | ||
| NM_001291449.1 | NP_001278378.1 | 2 | 271 | |||
| NM_172291.2 | NP_758495.1 | 2 | 271 | |||
| GenBank mRNA | AK054231.1 | - | 2 | 163 | ||
| AK144759.1 | - | 2 | 197 | |||
| AK163722.1 | - | 2 | 130 | |||
| BC017604.1 | - | 2 | 224 | |||
| BC024806.1 | - | 2 | 141 | 5361053 | ||
| BC027386.1 | - | 2 | 170 | |||
| BC028892.1 | - | 1 | 827 | |||

Foxred1
Genomic Map