siRNA Details
| Gene Symbol: Zfp759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172392.3 | NP_765980.3 | 4 | 419 | ||
| GenBank mRNA | AK030126.1 | - | 4 | 422 | ||
| BC028265.1 | - | 4 | 403 | 5365910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp759| Gene Symbol: Zfp759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172392.3 | NP_765980.3 | 4 | 419 | ||
| GenBank mRNA | AK030126.1 | - | 4 | 422 | ||
| BC028265.1 | - | 4 | 403 | 5365910 | ||