siRNA Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172405.3 | NP_765993.1 | 2 | 117 | ||
| GenBank mRNA | AK014420.1 | - | 2 | 143 | ||
| AK042001.1 | - | 2 | 126 | |||
| AK042339.1 | - | 2 | 126 | |||
| AK051816.1 | - | 2 | 118 | |||
| AK167540.1 | - | 2 | 148 | |||
| BC029845.1 | - | 2 | 122 | 4010744 | ||

Fam175a
Genomic Map