siRNA Details
| Gene Symbol: 9030612E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018547.1 | - | 1 | 370 | ||
| AK029173.1 | - | 1 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030612E09Rik| Gene Symbol: 9030612E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018547.1 | - | 1 | 370 | ||
| AK029173.1 | - | 1 | 365 | |||