siRNA Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172449.2 | NP_766037.2 | 25, 26 | 5994 | ||
| GenBank mRNA | AK051895.1 | - | 4, 5 | 1943 | ||
| BC055691.1 | - | 5, 6 | 745 | |||
| BC066146.1 | - | 4, 5 | 1005 | 30470084 | ||
| BC083181.1 | - | 7, 8 | 968 | |||

Bzrap1
Genomic Map