siRNA Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159527.1 | NP_001152999.1 | 2 | 235 | ||
| NM_172470.3 | NP_766058.3 | 2 | 235 | |||
| GenBank mRNA | AK015490.1 | - | 2 | 200 | ||
| AK029896.1 | - | 2 | 190 | |||
| AK031061.1 | - | 2 | 226 | |||
| AK053724.1 | - | 2 | 190 | |||
| AK083971.1 | - | 2 | 180 | |||
| AK173151.1 | - | 2 | 177 | |||
| BC056925.1 | - | 2 | 344 | 5704780 | ||

Wdr35
Genomic Map