siRNA Details
| Gene Symbol: Zfp180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045486.2 | NP_001038951.1 | 5 | 1083 | ||
| NM_001289634.1 | NP_001276563.1 | 6 | 1187 | |||
| NM_001289637.1 | NP_001276566.1 | 4 | 1002 | |||
| NM_001289640.1 | NP_001276569.1 | 5 | 1078 | |||
| NM_001289641.1 | NP_001276570.1 | 5 | 1043 | |||
| NM_172483.2 | NP_766071.1 | 3 | 887 | |||
| GenBank mRNA | AK083802.1 | - | 3 | 861 | ||
| AK141922.1 | - | 5 | 1036 | |||
| AK144892.1 | - | 5 | 1044 | |||
| AK145004.1 | - | 5 | 1048 | |||
| AK154182.1 | - | 5 | 1040 | |||
| AY166794.1 | - | 2 | 543 | |||
| BC063741.1 | - | 5 | 1021 | 6504558 | ||
| BC066115.1 | - | 5 | 1045 | 30619470 | ||
| BC085472.1 | - | 5 | 1056 | 6417473 | ||

Zfp180
Genomic Map