siRNA Details
| Gene Symbol: Zfp677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172486.2 | NP_766074.2 | 3 | 441 | ||
| GenBank mRNA | AK043955.1 | - | 3 | 443 | ||
| BB641850.1 | - | 3 | 443 | |||
| BC057947.1 | - | 3 | 419 | 5344769 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp677| Gene Symbol: Zfp677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172486.2 | NP_766074.2 | 3 | 441 | ||
| GenBank mRNA | AK043955.1 | - | 3 | 443 | ||
| BB641850.1 | - | 3 | 443 | |||
| BC057947.1 | - | 3 | 419 | 5344769 | ||