siRNA Details
| Gene Symbol: Tmem125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172383.3 | NP_759015.2 | 4 | 1296 | ||
| GenBank mRNA | AK050146.1 | - | 4 | 1300 | ||
| AK160304.1 | - | 2 | 951 | |||
| AK162829.1 | - | 911 | ||||
| AK162907.1 | - | 2 | 967 | |||
| BC039806.1 | - | 1 | 794 | 1516024 | ||

Tmem125
Genomic Map