siRNA Details
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172397.3 | NP_765985.1 | 5 | 469 | ||
GenBank mRNA | AK012581.1 | - | 5 | 443 | ||
AK032430.1 | - | 5 | 470 | |||
AK040486.1 | - | 5 | 470 | |||
AK049809.1 | - | 5 | 439 | |||
AK167349.1 | - | 5 | 439 | |||
AK169829.1 | - | 5 | 440 | |||
AK171794.1 | - | 5 | 748 | |||
AK187911.1 | - | 2 | 169 | |||
AK188791.1 | - | 2 | 168 | |||
BC068130.1 | - | 5 | 413 | 6413206 |
