siRNA Details
| Gene Symbol: Slc25a32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172402.3 | NP_765990.2 | 2 | 461 | ||
| GenBank mRNA | AK031187.1 | - | 2 | 266 | ||
| AK145178.1 | - | 2 | 421 | |||
| AK167501.1 | - | 2 | 271 | |||
| AK170643.1 | - | 2 | 430 | |||
| AK172029.1 | - | 2 | 429 | |||
| AK172597.1 | - | 2 | 419 | |||
| BB641480.1 | - | 2 | 462 | |||
| BC031874.1 | - | 2 | 395 | 5149984 | ||

Slc25a32
Genomic Map