siRNA Details
| Gene Symbol: Perm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172417.3 | NP_766005.2 | 2 | 1083 | ||
| GenBank mRNA | AK036660.1 | - | 2 | 1085 | ||
| AK040833.1 | - | 2 | 1077 | |||
| BC117871.1 | - | 1 | 858 | 40095536 | ||
| BC117872.1 | - | 1 | 858 | 40095540 | ||
| KF150176.1 | - | 1 | 750 | |||

Perm1
Genomic Map