siRNA Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172430.3 | NP_766018.3 | 2, 3 | 329 | ||
| GenBank mRNA | AK034183.1 | - | 3, 4 | 372 | ||
| AK038638.1 | - | 3, 4 | 374 | |||
| BC069832.1 | - | 2, 3 | 295 | 30431760 | ||
| BY769819.1 | - | 2, 3 | 331 | |||
| BY774616.1 | - | 2, 3 | 330 | |||

Sphkap
Genomic Map