siRNA Details
| Gene Symbol: TUBG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001070.4 | NP_001061.2 | 5, 6 | 885 | ||
| GenBank mRNA | AK313339.1 | - | 5, 6 | 554 | ||
| BC000619.2 | - | 5, 6 | 549 | 3345973 | ||
| BT019931.1 | - | 5, 6 | 487 | |||
| CR407642.1 | - | 5, 6 | 487 | |||
| M61764.1 | - | 5, 6 | 511 | |||

TUBG1
Genomic Map