siRNA Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172136.1 | NP_001165607.1 | 4 | 541 | ||
| NM_172456.3 | NP_766044.1 | 3 | 506 | |||
| GenBank mRNA | AK028887.1 | - | 3 | 470 | ||
| AK034654.1 | - | 4 | 503 | |||
| AK051300.1 | - | 4 | 505 | |||
| AK148426.1 | - | 3 | 487 | |||
| AK162566.1 | - | 3 | 469 | |||
| AK163555.1 | - | 3 | 470 | |||
| BC059920.1 | - | 4 | 484 | |||
| BC064730.1 | - | 4 | 556 | |||
| BC094589.1 | - | 2 | 1743 | |||
| BC119024.1 | - | 3 | 470 | 8733773 | ||
| BC144729.1 | - | 4 | 505 | |||

Exog
Genomic Map