siRNA Details
| Gene Symbol: Sned1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172463.4 | NP_766051.4 | 28 | 4112 | ||
| GenBank mRNA | AJ584850.1 | - | 28 | 4168 | ||
| AK032805.1 | - | 6 | 798 | |||
| AK139277.1 | - | 3 | 224 | |||
| BC145886.1 | - | 28 | 4112 | 40131189 | ||

Sned1
Genomic Map