siRNA Details
| Gene Symbol: Frmd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177843.1 | NP_001171314.1 | 3 | 755 | ||
| NM_172475.3 | NP_766063.3 | 3 | 385 | |||
| GenBank mRNA | AK043369.1 | - | 3 | 262 | ||
| AK078493.1 | - | 3 | 259 | |||
| AK082365.1 | - | 3 | 331 | |||
| BC058672.1 | - | 3 | 376 | 6830456 | ||
| BC082808.1 | - | 3 | 762 | |||

Frmd4a
Genomic Map