siRNA Details
| Gene Symbol: Zfp536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172385.2 | NP_759017.1 | 6 | 2821 | ||
| GenBank mRNA | AK030633.1 | - | 6 | 2822 | ||
| AK034508.1 | - | 6 | 2765 | |||
| AK042625.1 | - | 3 | 881 | |||
| AK122270.1 | - | 7 | 2828 | |||
| BC033594.1 | - | 6 | 2743 | 5367892 | ||

Zfp536
Genomic Map