siRNA Details
| Gene Symbol: Slc25a32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172402.3 | NP_765990.2 | 7 | 2167 | ||
| GenBank mRNA | AK011759.1 | - | 1 | 1084 | ||
| AK031187.1 | - | 7 | 1972 | |||
| AK145178.1 | - | 7 | 2127 | |||
| AK167501.1 | - | 7 | 1977 | |||
| AK170643.1 | - | 7 | 2136 | |||
| AK172029.1 | - | 7 | 2134 | |||
| AK219382.1 | - | 1 | 296 | |||
| BC031874.1 | - | 7 | 2101 | 5149984 | ||

Slc25a32
Genomic Map