siRNA Details
| Gene Symbol: 2310007B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159940.1 | NP_001153412.1 | 6 | 1994 | ||
| NM_172411.4 | NP_765999.3 | 6 | 1985 | |||
| GenBank mRNA | AK009193.1 | - | 6 | 1994 | ||
| AK081300.1 | - | 5 | 1873 | |||
| BC089525.1 | - | 6 | 1985 | 30315016 | ||

2310007B03Rik
Genomic Map