siRNA Details
| Gene Symbol: Zc2hc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172414.4 | NP_766002.2 | 3 | 4139 | ||
| GenBank mRNA | AK036602.1 | - | 3 | 4077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc2hc1c| Gene Symbol: Zc2hc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172414.4 | NP_766002.2 | 3 | 4139 | ||
| GenBank mRNA | AK036602.1 | - | 3 | 4077 | ||