siRNA Details
| Gene Symbol: Ostm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172416.3 | NP_766004.1 | 6 | 2380 | ||
| GenBank mRNA | AF533890.1 | - | 6 | 2379 | ||
| AK004546.1 | - | 6 | 2357 | |||
| AK044897.1 | - | 6 | 2378 | |||
| AK050513.1 | - | 6 | 2354 | |||
| BC057635.1 | - | 6 | 2319 | 4910872 | ||

Ostm1
Genomic Map