siRNA Details
| Gene Symbol: Perm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172417.3 | NP_766005.2 | 4 | 3606 | ||
| GenBank mRNA | AK009747.2 | - | 1 | 877 | ||
| AK036660.1 | - | 4 | 3608 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Perm1| Gene Symbol: Perm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172417.3 | NP_766005.2 | 4 | 3606 | ||
| GenBank mRNA | AK009747.2 | - | 1 | 877 | ||
| AK036660.1 | - | 4 | 3608 | |||